4-Amino-2-chlorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
3-Chloro-4-cyanoaniline
A2784-25G
A2784-5G
TCA2784-25GEA
359
USD
TCA2784-25G
TCA2784-5G
4-Amino-2-chlorobenzonitrile ≥98.0% (by GC)
4-Amino-2-chlorobenzonitrile
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD00035926 CAS Number: 20925-27-3 EINECS: 244-114-0 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...