3,4,5-Trimethoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

T3244-25G T3244-5G
TCT3244-25GEA 333 USD
TCT3244-25G TCT3244-5G
3,4,5-Trimethoxybenzonitrile ≥98.0% (by GC)
3,4,5-Trimethoxybenzonitrile
Formula: C₁₀H₁₁NO₃
MW: 193.2 g/mol
Boiling Pt: 185°C
MDL Number: MFCD00001803
CAS Number: 1885-35-4
EINECS: 217-550-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 96.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR