2,3,4-Trimethoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

T3285-25G T3285-5G
TCT3285-25GEA 195 USD
TCT3285-25G TCT3285-5G
2,3,4-Trimethoxybenzonitrile ≥98.0% (by GC)
2,3,4-Trimethoxybenzonitrile
Formula: C₁₀H₁₁NO₃
MW: 193.2 g/mol
Boiling Pt: 167°C
MDL Number: MFCD00001785
CAS Number: 43020-38-8
EINECS: 256-049-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR