2,3-Dimethoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

D4853-25G D4853-5G
TCD4853-25GEA 207 USD
TCD4853-25G TCD4853-5G
2,3-Dimethoxybenzonitrile ≥98.0% (by GC)
2,3-Dimethoxybenzonitrile
Formula: C₉H₉NO₂
MDL Number: MFCD00001784
CAS Number: 5653-62-3
EINECS: 227-097-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR