2-Bromo-4-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

B4873-1G B4873-5G
TCB4873-1GEA 43 USD
TCB4873-1G TCB4873-5G
2-Bromo-4-nitroaniline ≥98.0% (by GC)
2-Bromo-4-nitroaniline
Formula: C₆H₅BrN₂O₂
MDL Number: MFCD00025152
CAS Number: 13296-94-1
EINECS: 236-318-3

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR