Propyne ∼5% ∼1 mol/L in THF

Supplier: TCI America
Danger

P2295-100ML P2295-500ML
TCP2295-100MLEA 77.19 USD
TCP2295-100ML TCP2295-500ML
Propyne ∼5% ∼1 mol/L in THF
Propyne
Formula: C₃H₄
Storage Temperature: Refrigerator
MDL Number: MFCD00036235
CAS Number: 74-99-7
EINECS: 200-828-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Concentration (HNMR) 4.5 to 6.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR