3-Methyl-1-butene ∼16.5% ∼2 mol/L in DMF

Supplier: TCI America
Danger

M2564-100ML M2564-500ML
TCM2564-100MLEA 107.8 USD
TCM2564-100ML TCM2564-500ML
3-Methyl-1-butene ∼16.5% ∼2 mol/L in DMF
3-Methyl-1-butene
Formula: C₅H₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD00008937
CAS Number: 563-45-1
EINECS: 209-249-1
UN: 1992
ADR: 3,I

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 16.0% to 18.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR