2-Methylpropene ∼8% in dichloromethane

Supplier: TCI America
Danger

I0909-100ML I0909-500ML
TCI0909-100MLEA 64.16 USD
TCI0909-100ML TCI0909-500ML
2-Methylpropene ∼8% in dichloromethane
2-Methylpropene
Formula: C₄H₈
Storage Temperature: Refrigerator
MDL Number: MFCD00008898
CAS Number: 115-11-7
EINECS: 204-066-3
UN: 1593
ADR: 6.1,III
Merck Index: 14,05141

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 8.0 to 9.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR