1-Butene ∼10% in hexane

Supplier: TCI America
Danger

B4410-100ML B4410-500ML
TCB4410-100MLEA 50 USD
TCB4410-100ML TCB4410-500ML
1-Butene ∼10% in hexane
1-Butene
Formula: C₄H₈
Storage Temperature: Refrigerator
MDL Number: MFCD00009383
CAS Number: 106-98-9
EINECS: 203-449-2
UN: 1993
ADR: 3,II
Merck Index: 14,01519

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 10.0 to 12.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR