Chloroethane ∼15% ∼2 mol/L in THF

Supplier: TCI America
Danger

Synonyms: Ethyl chloride

C2883-100ML C2883-500ML
TCC2883-100MLEA 121 USD
TCC2883-100ML TCC2883-500ML
Chloroethane ∼15% ∼2 mol/L in THF
Chloroethane
Formula: C₂H₅Cl
MW: 64.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00000961
CAS Number: 75-00-3
EINECS: 200-830-5
UN: 1993
ADR: 3,II
Merck Index: 14,03782

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 14.5 to 15.5 w/w%
Water max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR