1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2R)-2-(dimethylamino)cyclohexyl)urea 98%

Supplier: Ambeed
Warning

A775987-100MG A775987-250MG
77618-124EA 69.01 USD
77618-124 77618-126
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2R)-2-(dimethylamino)cyclohexyl)urea 98%
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2R)-2-(dimethylamino)cyclohexyl)urea

Precaución: for Research Use Only

Formula: C₁₇H₂₁F₆N₃O
MW: 397.3586 g/mol
CAS Number: 820242-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR