2,2'-bis[(R)-α-(dimethylamino)benzyl]ferrocene 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-1,1'-Bis(dicyclohexylphosphino)-2,2'-bis[(R)-α-(dimethylamino)benzyl]ferrocene, (S,S)-(+)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(dicyclohexylphosphino)ferrocene

A735381-100MG A735381-250MG
77618-012EA 98.88 USD
77618-012 77618-014
2,2'-bis[(R)-α-(dimethylamino)benzyl]ferrocene 98%
2,2'-bis[(R)-α-(dimethylamino)benzyl]ferrocene

Precaución: for Research Use Only

Formula: C₅₂H₇₄FeN₂P₂
MW: 844.95 g/mol
MDL Number: MFCD04117702
CAS Number: 494227-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR