12-Aminododecan-1-ol 98%

Supplier: Ambeed
Warning

A625019-1G A625019-5G
77617-634EA 116.39 USD
77617-634 77747-288
12-Aminododecan-1-ol 98%
12-Aminododecan-1-ol

Precaución: for Research Use Only

Formula: C₁₂H₂₇NO
MDL Number: MFCD01851111
CAS Number: 67107-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR