(2R,4R)-1-(tert-Butoxycarbonyl)-2-methylpiperidine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A544153-250MG A544153-100MG A544153-1G
77617-322EA 697.31 USD
77617-322 77617-320 77686-402
(2R,4R)-1-(tert-Butoxycarbonyl)-2-methylpiperidine-4-carboxylic acid 97%
(2R,4R)-1-(tert-Butoxycarbonyl)-2-methylpiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD29058646
CAS Number: 1932555-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR