N-(4-(Acridin-9-ylamino)-3-methoxyphenyl)ethanesulfonamide methanesulfonate 97%

Supplier: Ambeed
Warning

A354235-1MG A354235-10MG A354235-5MG
77616-548EA 59.74 USD
77616-548 77748-702 77748-703
N-(4-(Acridin-9-ylamino)-3-methoxyphenyl)ethanesulfonamide methanesulfonate 97%
N-(4-(Acridin-9-ylamino)-3-methoxyphenyl)ethanesulfonamide methanesulfonate

Precaución: for Research Use Only

Formula: C₂₃H₂₅N₃O₆S₂
CAS Number: 59988-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR