3-Hydrazinobenzonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 3-Cyanophenylhydrazine

A302316-5G A302316-10G A302316-1G
77616-308EA 88.58 USD
77616-308 77748-584 77748-585
3-Hydrazinobenzonitrile 95%
3-Hydrazinobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.151 g/mol
MDL Number: MFCD03030048
CAS Number: 17672-26-3
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR