1,9-Dichlorononane ≥98.0% (by GC)

Supplier: TCI America
Warning

D5116-25G D5116-5G
76219-160EA 110 USD
76219-160 76219-162
1,9-Dichlorononane ≥98.0% (by GC)
1,9-Dichlorononane
Formula: C₉H₁₈Cl₂
Boiling Pt: 150°C
MDL Number: MFCD00001024
CAS Number: 821-99-8
EINECS: 212-484-2
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR