1,3-Diiodobicyclo[1.1.1]pentane 95%

Supplier: Ambeed
Warning

A263071-100MG A263071-1G A263071-250MG
77616-106EA 221.45 USD
77616-106 77746-446 77746-447
1,3-Diiodobicyclo[1.1.1]pentane 95%
1,3-Diiodobicyclo[1.1.1]pentane

Precaución: for Research Use Only

Formula: C₅H₆I₂
MDL Number: MFCD29037325
CAS Number: 105542-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR