(S)-1,4-Diazabicyclo[4.3.0]nonane 95%

Supplier: Ambeed
Danger

A242053-1G A242053-250MG
77615-994EA 179.22 USD
77615-994 77746-364
(S)-1,4-Diazabicyclo[4.3.0]nonane 95%
(S)-1,4-Diazabicyclo[4.3.0]nonane

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
MW: 126.20 g/mol
MDL Number: MFCD03787926
CAS Number: 93643-24-4
UN: 3267
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR