Pelargonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Nonanoic acid, Pelargonic Acid

P0952-25ML P0952-500ML
TCP0952-25MLEA 21 USD
TCP0952-25ML TCP0952-500ML
Pelargonic acid ≥98.0% (by titrimetric analysis)
Pelargonic acid
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 254°C
Flash Pt: 140°C
MDL Number: MFCD00004433
CAS Number: 112-05-0
EINECS: 203-931-2
UN: 3265
ADR: 8,III
Merck Index: 14,07070

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR