2-(4-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride 98%

Supplier: Ambeed
Warning

A883623-1MG A883623-10MG A883623-5MG A883623-100MG A883623-50MG
77614-588EA 84.46 USD
77614-588 77686-924 77686-925 77900-622 77900-859
2-(4-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride 98%
2-(4-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃Cl₂N₃
MDL Number: MFCD09878232
CAS Number: 36376-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR