Bis(NHS)PEG₉ 98%

Supplier: Ambeed
Warning

Synonyms: Bis(NHS)PEG9, 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioic acid di(N-succinimidyl) ester, Di(N-succinimidyl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate, 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioicacid di(N-succinimidyl) ester, Bis(2,5-dioxopyrrolidin-1-yl) 4,7,10,13,16,19,22,25,28-nonaoxahentriacontanedioate, Di(N-succinimidyl) 4,7,10,13,16,19,22,25,28-Nonaoxahentriacontanedioate, Bis-dPEG®9-NHS ester

A868622-250MG A868622-100MG A868622-1G
77614-568EA 154.5 USD
77614-568 77686-908 77686-909
Bis(NHS)PEG₉ 98%
Bis(NHS)PEG9

Precaución: for Research Use Only

Formula: C₃₀H₄₈N₂O₁₇
MDL Number: MFCD11041131
CAS Number: 1008402-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR