tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3,3-difluoro-1-(hydroxymethyl)cyclobutyl)carbamate

A594641-1G A594641-250MG
77613-846EA 264.71 USD
77613-846 77686-513
tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate 97%
tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₇F₂NO₃
MW: 237.244 g/mol
MDL Number: MFCD22566207
CAS Number: 1232365-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR