3,4-Dimethoxyphenethyl bromide 95%

Supplier: Ambeed
Danger

A554042-1G A554042-10G A554042-25G
77613-744EA 55.62 USD
77613-744 77886-366 77886-367
3,4-Dimethoxyphenethyl bromide 95%
3,4-Dimethoxyphenethyl bromide

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₂
MDL Number: MFCD07369769
CAS Number: 40173-90-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR