4-Bromoquinolin-7-ol 95%

Supplier: Ambeed
Danger

A543407-100MG A543407-1G
77613-704EA 66.39 USD
77613-704 77886-356
4-Bromoquinolin-7-ol 95%
4-Bromoquinolin-7-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD09261121
CAS Number: 181950-60-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR