TG-89 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(tert-Butyl)-3-((5-methyl-2-((3-(morpholinomethyl)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide
A538821-1MG
A538821-5MG
77613-688EA
37.08
USD
77613-688
77686-388
TG-89 95+%
TG-89
Precaución: for Research Use Only
Formula:
C₂₆H₃₄N₆O₃S MW: 510.7 g/mol |
MDL Number:
MFCD16620524 CAS Number: 936091-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...