4-Methoxy-2-(((5-methoxy-1H-benzo[d]imidazol-2-yl)sulfinyl)methyl)-3,5-dimethylpyridine 1-oxide 98%, Ambeed.Inc

Supplier: Ambeed
Danger

A527576-1MG A527576-5MG A527576-100MG A527576-1G A527576-250MG
77613-658EA 15.45 USD
77613-658 77684-365 77886-329 77886-330 77886-331
4-Methoxy-2-(((5-methoxy-1H-benzo[d]imidazol-2-yl)sulfinyl)methyl)-3,5-dimethylpyridine 1-oxide 98%, Ambeed.Inc
4-Methoxy-2-(((5-methoxy-1H-benzo[d]imidazol-2-yl)sulfinyl)methyl)-3,5-dimethylpyridine 1-oxide

Precaución: For research use only.

Formula: C₁₇H₁₉N₃O₄S
MDL Number: MFCD08274751
CAS Number: 176219-04-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR