4-(3-Aminopropyl)aniline 95%

Supplier: Ambeed
Warning

A519522-250MG A519522-100MG A519522-1G
77613-632EA 136.99 USD
77613-632 77673-382 77673-384
4-(3-Aminopropyl)aniline 95%
4-(3-Aminopropyl)aniline

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MDL Number: MFCD09744485
CAS Number: 61798-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR