tert-Butyl (cis-3-amino-3-methylcyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

A394029-250MG A394029-100MG A394029-1G
77613-194EA 341.96 USD
77613-194 77686-088 77686-089
tert-Butyl (cis-3-amino-3-methylcyclobutyl)carbamate 97%
tert-Butyl (cis-3-amino-3-methylcyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD23106027
CAS Number: 1408076-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR