(S)-β-(4-Chlorophenyl)alaninol 95%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Amino-3-(4-chlorophenyl)-1-propanol, (S)-3-Amino-3-(4-chlorophenyl)propan-1-ol

A362006-5G A362006-100MG
77613-062EA 205.2 USD
77613-062 77686-024
(S)-β-(4-Chlorophenyl)alaninol 95%
(S)-β-(4-Chlorophenyl)alaninol

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO
MDL Number: MFCD07784069
CAS Number: 886061-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR