1,1,5,5-Tetrafluoro-2,4-pentanedione 98%

Supplier: Ambeed
Danger

A316276-1G A316276-250MG A316276-5G
77612-824EA 152.89 USD
77612-824 77612-826 77883-858
1,1,5,5-Tetrafluoro-2,4-pentanedione 98%
1,1,5,5-Tetrafluoro-2,4-pentanedione

Precaución: for Research Use Only

Formula: C₅H₄F₄O₂
MDL Number: MFCD03419795
CAS Number: 70086-62-3
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR