L-tert-Leucine methyl ester 98%

Supplier: Ambeed
Danger

Synonyms: (S)-Methyl 2-amino-3,3-dimethylbutanoate, (S)-2-Amino-3,3-dimethylbutyric Acid Methyl Ester, H-Tle-OMe, Methyl (S)-2-Amino-3,3-dimethylbutyrate

A960174-1G A960174-250MG A960174-5G
77603-358EA 102.5 USD
77603-358 77603-360 77603-362
L-tert-Leucine methyl ester 98%
L-tert-Leucine methyl ester

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MDL Number: MFCD00137409
CAS Number: 63038-26-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR