1,3-Dihydroxyacetone ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Dihydroxyacetone
D5818-25G
D5818-5G
TCD5818-25GEA
65
USD
TCD5818-25G
TCD5818-5G
1,3-Dihydroxyacetone ≥97.0% (by GC)
1,3-Dihydroxyacetone
Formula:
C₃H₆O₃ MW: 90.08 g/mol |
MDL Number:
MFCD00004670 CAS Number: 96-26-4 EINECS: 202-494-5 Merck Index: 14,03182 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...