m-PEG10-acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,5,8,11,14,17,20,23,26,29-Decaoxadotriacontan-32-oic acid

A750781-1G A750781-250MG
77602-918EA 282.9 USD
77602-918 77602-920
m-PEG10-acid 95%
m-PEG10-acid

Precaución: for Research Use Only

Formula: C₂₂H₄₄O₁₂
MW: 500.578 g/mol
MDL Number: MFCD27977513
CAS Number: 2409969-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR