Bis(2-methoxyphenyl)phenylphosphine 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2-methoxyphenyl)(phenyl)phosphine

A714292-1G A714292-5G A714292-10G
77602-824EA 20.5 USD
77602-824 77602-826 77878-559
Bis(2-methoxyphenyl)phenylphosphine 97%
Bis(2-methoxyphenyl)phenylphosphine

Precaución: for Research Use Only

Formula: C₂₀H₁₉O₂P
MDL Number: MFCD00061478
CAS Number: 36802-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR