β-Nicotinamide adenine dinucleotide sodium salt 98%

Supplier: Ambeed
Warning

A554303-1G A554303-25G A554303-5G A554303-100G
77602-352EA 11.28 USD
77602-352 77602-354 77602-356 77878-345
β-Nicotinamide adenine dinucleotide sodium salt 98%
β-Nicotinamide adenine dinucleotide sodium salt

Precaución: for Research Use Only

Formula: C₂₁H₂₆N₇NaO₁₄P₂
MDL Number: MFCD00148917
CAS Number: 20111-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR