(S)-2-(3,4-Dihydroxyphenyl)-5-hydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one 98%

Supplier: Ambeed
Warning

A509884-1MG A509884-10MG A509884-5MG
77602-180EA 23.58 USD
77602-180 77627-006 77627-008
(S)-2-(3,4-Dihydroxyphenyl)-5-hydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one 98%
(S)-2-(3,4-Dihydroxyphenyl)-5-hydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

Precaución: for Research Use Only

Formula: C₂₁H₂₂O₁₁
MDL Number: MFCD18632528
CAS Number: 38965-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR