n-Dodecanoylsucrose 97%

Supplier: Ambeed
Warning

A481375-100MG A481375-1G A481375-250MG
77602-082EA 29.73 USD
77602-082 77602-084 77669-160
n-Dodecanoylsucrose 97%
n-Dodecanoylsucrose

Precaución: for Research Use Only

Formula: C₂₄H₄₄O₁₂
MDL Number: MFCD00077947
CAS Number: 25339-99-5
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR