Dipentaerythritol Hexakis(3-mercaptopropionate) 93%

Supplier: Ambeed
Warning

Synonyms: DiPETMP, Dipentaerythritol hexa(3-mercaptopropionate)

A422045-500G A422045-100G
77601-822EA 200.9 USD
77601-822 77601-820
Dipentaerythritol Hexakis(3-mercaptopropionate) 93%
Dipentaerythritol Hexakis(3-mercaptopropionate)

Precaución: for Research Use Only

Formula: C₂₈H₄₆O₁₃S₆
MW: 783.02 g/mol
MDL Number: MFCD00128217
CAS Number: 25359-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR