Aminoacetonitrile 97%

Supplier: Ambeed
Danger

A396915-1G A396915-25G A396915-5G A396915-10G
77601-750EA 36.9 USD
77601-750 77601-752 77601-754 77626-810
Aminoacetonitrile 97%
Aminoacetonitrile

Precaución: for Research Use Only

Formula: C₂H₄N₂
MW: 56.0666 g/mol
MDL Number: MFCD00078890
CAS Number: 540-61-4
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR