Migalastat hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: AT1001, AT-1001, GR181413A, 1,5-Dideoxy-1,5-imino-D-galactitol, 1-Deoxygalactonojirimycin, 1-Deoxygalactostatin

A312646-1MG A312646-5MG
77601-408EA 24.6 USD
77601-408 77626-560
Migalastat hydrochloride 98%
Migalastat hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₄·HCl
MW: 199.6 g/mol
MDL Number: MFCD00269962
CAS Number: 75172-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR