trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid (CDTA) 98%

Supplier: Ambeed
Warning

Synonyms: trans-Cyclohexane-1,2-dinitrilotetraacetic acid, 2,2',2'',2'''-((1R,2R)-rel-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid, trans-Cyclohex-1,2-ylenediaminetetra(acetic acid), CDTA, CyDTA, trans-Cyclohex-1,2-ylenediaminetetra acetic acid

A226407-1G A226407-5G A226407-25G
77600-814EA 9.23 USD
77600-814 77600-816 77626-164
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid (CDTA) 98%
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid (CDTA)

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂O₈
MDL Number: MFCD00003845
CAS Number: 13291-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR