Tris(2,4,6-trimethylphenyl)phosphine 98%

Supplier: Ambeed
Warning

Synonyms: Nalpha-Acetyl-D-arginine, Trimesitylphosphine

A190366-1G A190366-250MG A190366-10G A190366-5G
77600-548EA 69.7 USD
77600-548 77600-550 77881-420 77881-421
Tris(2,4,6-trimethylphenyl)phosphine 98%
Tris(2,4,6-trimethylphenyl)phosphine

Precaución: for Research Use Only

Formula: C₂₇H₃₃P
MDL Number: MFCD00014912
CAS Number: 23897-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR