3-Hydroxy-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

A1371212-100MG A1371212-250MG A1371212-1G
77599-994EA 59.45 USD
77599-994 77599-996 77621-888
3-Hydroxy-2-methylpropanoic acid 95%
3-Hydroxy-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.1045 g/mol
MDL Number: MFCD11975300
CAS Number: 2068-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR