1,4-Dibromopentane ≥95.0% (by GC)

Supplier: TCI America
Warning

D1408-25G D1408-5G
76218-972EA 63.09 USD
76218-972 76218-974
1,4-Dibromopentane ≥95.0% (by GC)
1,4-Dibromopentane
Formula: BrH₂C(CH₂)₂BrCHCH₃
MW: 229.94 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00000153
CAS Number: 626-87-9
EINECS: 210-967-2

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR