Oleyl alcohol 80% -85%

Supplier: Ambeed
Warning

A132058-100G A132058-25G
77599-902EA 26.65 USD
77599-902 77666-900
Oleyl alcohol 80% -85%
Oleyl alcohol

Precaución: for Research Use Only

Formula: C₁₈H₃₆O
MW: 268.48 g/mol
MDL Number: MFCD00002993
CAS Number: 143-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR