(R)-2-((5-(1,2-Dithiolan-3-yl)pentanoyl)oxy)-N,N,N-trimethylethan-1-aminium chloride 98%

Supplier: Ambeed
Danger

A1227171-100MG A1227171-250MG A1227171-50MG
77599-782EA 190.65 USD
77599-782 77621-152 77621-154
(R)-2-((5-(1,2-Dithiolan-3-yl)pentanoyl)oxy)-N,N,N-trimethylethan-1-aminium chloride 98%
(R)-2-((5-(1,2-Dithiolan-3-yl)pentanoyl)oxy)-N,N,N-trimethylethan-1-aminium chloride

Precaución: for Research Use Only

Formula: C₁₃H₂₆ClNO₂S₂
CAS Number: 1808266-58-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR