Z-Pro-Pro-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-((S)-1-((Benzyloxy)carbonyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxylic acid

A101916-1G A101916-5G
77599-490EA 48.18 USD
77599-490 77666-478
Z-Pro-Pro-OH 97%
Z-Pro-Pro-OH

Precaución: for Research Use Only

Formula: C₁₈H₂₂N₂O₅
MW: 346.383 g/mol
MDL Number: MFCD00077035
CAS Number: 7360-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR