10-Methyl-9-phenylacridinium perchlorate 98%

Supplier: Ambeed
Danger

A989763-250MG A989763-1G A989763-5G
77599-404EA 43.05 USD
77599-404 77599-402 77623-366
10-Methyl-9-phenylacridinium perchlorate 98%
10-Methyl-9-phenylacridinium perchlorate

Precaución: for Research Use Only

Formula: C₂₀H₁₆ClNO₄
MDL Number: MFCD09038537
CAS Number: 36519-61-6
UN: 1479
ADR: 5.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR