(S)-N-(4-(((2S,5R)-5-((R)-Hydroxy(phenyl)methyl)pyrrolidin-2-yl)methyl)phenyl)-4-oxo-4,6,7,8-tetrahydropyrrolo[1,2-a]pyrimidine-6-carboxamide 97%
Supplier: Ambeed
|
Warning
|
A846150-1MG
A846150-10MG
A846150-5MG
77599-172EA
36.9
USD
77599-172
77623-044
77623-046
(S)-N-(4-(((2S,5R)-5-((R)-Hydroxy(phenyl)methyl)pyrrolidin-2-yl)methyl)phenyl)-4-oxo-4,6,7,8-tetrahydropyrrolo[1,2-a]pyrimidine-6-carboxamide 97%
(S)-N-(4-(((2S,5R)-5-((R)-Hydroxy(phenyl)methyl)pyrrolidin-2-yl)methyl)phenyl)-4-oxo-4,6,7,8-tetrahydropyrrolo[1,2-a]pyrimidine-6-carboxamide
Precaución: for Research Use Only
Formula:
C₂₆H₂₈N₄O₃ MW: 444.5255 g/mol |
MDL Number:
MFCD28502057 CAS Number: 1190389-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...